Learn More
Sarstedt Inc 0.5ML SCMTUBE CBS WB NC PK/250
Supplier:  Sarstedt Inc 72.730.309
Encompass_Preferred
Screw Cap Micro Tube, 0.5 ml, PP, with smear edge, with knurls, without closure, with printed writing space and graduation, 250 pcs./bag.
Catalog No. NC1532986