Learn More
Sarstedt Inc 0.5ML SCMTUBE NOKNURL S/PK1000
Catalog No. NC9547075 Shop All Sarstedt Inc. Products
Encompass_Preferred
0.5ml Screw Cap Micro Tube Conical Bottom Skirted No Knurls O-Ring Cap Assembled Sterile
Catalog No. NC9547075 Supplier Sarstedt Inc Supplier No. 72.785.005
0.5ml Screw Cap Micro Tube Conical Bottom Skirted No Knurls O-Ring Cap Assembled Sterile