Learn More
Andwin Scientific 0.5ML SCREW CAP MIC/TB PK/500
SDP
Catalog No. NC9030993
Encompass_Preferred
The Screw-Cap Microcentrifuge Tubes are superior laboratory supplies for safe sample storage and handling. The tubes are free of detectable DNase and RNase and are autoclavable and freezable. This product is freestanding and is pre-sterilized by gamm
Catalog No. NC9030993 Supplier Andwin Scientific Supplier No. 89004286
The Screw-Cap Microcentrifuge Tubes are superior laboratory supplies for safe sample storage and handling. The tubes are free of detectable DNase and RNase and are autoclavable and freezable. This product is freestanding and is pre-sterilized by gamm