Learn More
USA Scientific Inc 0.5mL screw cap tube, wht
Catalog No. 502744932
Encompass
0.5 mL self-standing microcentrifuge tube, white, with integrated TPE cap, 500/pack
Catalog No. 50-274-4932 Supplier USA Scientific Inc Supplier No. 14059308
0.5 mL self-standing microcentrifuge tube, white, with integrated TPE cap, 500/pack