Learn More
CELLTREAT Scientific Products 0.5mL Screw Top Micro Tube and Cap, Self-Standing, Grip Band, Green Grip Cap With Integrated O-Ring, Sterile, 500/CS
SDP
Catalog No. 501535108
Encompass_Preferred
CELLTREAT Scientific Products 0.5mL Screw Top Micro Tube and Cap. High temperature resistivity range--shatter/crack resistant and ideal for boiling. Graduated tubes have markings every 100¿L for easy volume identification. Easy 3/4 turn to open and close. Sterile Assemblies packed in self-standing re-sealable bags offer a unique storage solution. Premium Grip Cap features molded TPE sealing rings which ensure O-ring will never slip out of place; striped TPE grips on cap edge enable secure handling under all conditions. Grip band tubes feature ribs that provide exceptional gripping surface and lessen movement during centrifugation. Temperature range of -196¿C to 121¿C (not for immersion in liquid nitrogen). Max RCF: 20,000g. Tubes manufactured from virgin polypropylene and are autoclavable. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 50/re-sealable bag, 500/case.
Catalog No. 50-153-5108 Supplier CELLTREAT Scientific Products Supplier No. 230810
CELLTREAT Scientific Products 0.5mL Screw Top Micro Tube and Cap. High temperature resistivity range--shatter/crack resistant and ideal for boiling. Graduated tubes have markings every 100¿L for easy volume identification. Easy 3/4 turn to open and close. Sterile Assemblies packed in self-standing re-sealable bags offer a unique storage solution. Premium Grip Cap features molded TPE sealing rings which ensure O-ring will never slip out of place; striped TPE grips on cap edge enable secure handling under all conditions. Grip band tubes feature ribs that provide exceptional gripping surface and lessen movement during centrifugation. Temperature range of -196¿C to 121¿C (not for immersion in liquid nitrogen). Max RCF: 20,000g. Tubes manufactured from virgin polypropylene and are autoclavable. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 50/re-sealable bag, 500/case.