Learn More
Chemglass Life Sciences Chemglass Life Sciences 0.5 mL Self-Standing Sample Vials, External Thread, Natural Caps, Sterile - CGN-4764-001N
SDP
Catalog No. 502733362
Encompass_Preferred
Sterile 0.5 mL self-standing sample vials with external thread and a sealing ring. Supplied in nested packaging and sterilized by electron beam to support sensitive laboratory workflows.
  • Capacity: 0.5 mL.
  • Vial type: Self-standing, round bottom.
  • Thread type: External thread.
  • Cap color: Natural color caps (inventory supplied in natural color).
  • Sealing: With sealing ring for secure closure.
  • Sterility: Sterile; electron beam sterilization (SAL = 10^-6).
  • Pack quantities: 50 per pack, 500 per box, 2000 per case.
  • Cleanliness: 100,000-level clean workshop production; DNase-free, RNase-free, pyrogen-free, endotoxin-free.
  • Packaging: Nest product line (nested packaging for handling and storage).
Catalog No. 50-273-3362 Supplier Chemglass Life Sciences Supplier No. CGN4764001N
Sterile 0.5 mL self-standing sample vials with external thread and a sealing ring. Supplied in nested packaging and sterilized by electron beam to support sensitive laboratory workflows.
  • Capacity: 0.5 mL.
  • Vial type: Self-standing, round bottom.
  • Thread type: External thread.
  • Cap color: Natural color caps (inventory supplied in natural color).
  • Sealing: With sealing ring for secure closure.
  • Sterility: Sterile; electron beam sterilization (SAL = 10^-6).
  • Pack quantities: 50 per pack, 500 per box, 2000 per case.
  • Cleanliness: 100,000-level clean workshop production; DNase-free, RNase-free, pyrogen-free, endotoxin-free.
  • Packaging: Nest product line (nested packaging for handling and storage).