Learn More
Chemglass Life Sciences Chemglass 0.5 mL Self-Standing Sample Vials, External Thread, Hinged Cap with Sealing Ring (CGN-4764-011N)
SDP
Catalog No. 502733358
Encompass_Preferred
Chemglass 0.5 mL self-standing sample vials (CGN-4764-011N) feature an external thread and hinged cap with a sealing ring. These sterile, nested vials are designed for safe, small-volume sample storage and handling in laboratory settings.
  • Capacity: 0.5 mL.
  • Hinged cap with sealing ring for a secure closure.
  • Natural color caps; photo may show blue caps.
  • Pack sizes: 50 per pack, 500 per box, 2000 per case.
  • Sterilized by electron beam; SAL=10-6.
  • External thread and self-standing round-bottom design.
  • Manufactured in 100,000-level clean workshop production.
  • DNase-free, RNase-free, pyrogen-free, and endotoxin-free.
Catalog No. 50-273-3358 Supplier Chemglass Life Sciences Supplier No. CGN4764011N
Chemglass 0.5 mL self-standing sample vials (CGN-4764-011N) feature an external thread and hinged cap with a sealing ring. These sterile, nested vials are designed for safe, small-volume sample storage and handling in laboratory settings.
  • Capacity: 0.5 mL.
  • Hinged cap with sealing ring for a secure closure.
  • Natural color caps; photo may show blue caps.
  • Pack sizes: 50 per pack, 500 per box, 2000 per case.
  • Sterilized by electron beam; SAL=10-6.
  • External thread and self-standing round-bottom design.
  • Manufactured in 100,000-level clean workshop production.
  • DNase-free, RNase-free, pyrogen-free, and endotoxin-free.