Learn More
SAI Infusion Technologies 500ul Na Citrate Blood Tubes (3.2%) with Flip Cap
SDP
Catalog No. NC1583707
Encompass
500ul 3.2% Sodium Citrate Blood Tube with attached Flip Cap. Ideal for collection of 500ul of blood. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/box. Product is for laboratory animal research only.
Catalog No. NC1583707 Supplier SAI Infusion Technologies Supplier No. PMTPC0.5
500ul 3.2% Sodium Citrate Blood Tube with attached Flip Cap. Ideal for collection of 500ul of blood. Slim Tube and Cap profiles ensure a perfect fit in standard racks and centrifuges. 100/box. Product is for laboratory animal research only.