Learn More
USA Scientific Inc 0.5 mL TempAssure PCR tube, attached flat cap, mixed
Catalog No. NC9776010
Encompass
0.5 mL TempAssure PCR tube, attached flat cap, mixed colors. DNase, RNase, DNA, PCR inhibitor, and pyrogen free. 1000/bag.
Catalog No. NC9776010 Supplier USA Scientific Inc Supplier No. 14058108
0.5 mL TempAssure PCR tube, attached flat cap, mixed colors. DNase, RNase, DNA, PCR inhibitor, and pyrogen free. 1000/bag.