Learn More
Alkali Scientific Qwik-Snap™ 0.5mL Microtube with Cap, Locktop Style Cap, 1000/cs
SDP
Catalog No. 502389075
Encompass_Preferred
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 1,000 clear 0.5mL centrifuge tubes.
Catalog No. 50-238-9075 Supplier Alkali Scientific Supplier No. C3050
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 1,000 clear 0.5mL centrifuge tubes.