Learn More
Ace Glass, Inc. 0-5mm high vacuum Easy Action stopcock with Tef-Cap O-ring
SDP
Catalog No. 50976143
Encompass_Preferred
0-5mm high vacuum Easy Action stopcock with Tef-Cap O-ring seals eliminate exposure to corrosive materials. For use to at least 13.6 Kg internal pressure. An ultimate vacuum of 10-7 can be realized with the standard three seals. Smooth acting valve permits fine adjustment of the opening. Front ring seal makes a positive closure against a precision-formed heavy glass seal. NOT bakeable. O-Rings are NOT replaceable. Reference marks on the stopcock body aid in repetitive setting. Replacement plugs with O-Rings sold separately.
Catalog No. 50-976-143 Supplier Ace Glass, Inc. Supplier No. 818905
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
0-5mm high vacuum Easy Action stopcock with Tef-Cap O-ring seals eliminate exposure to corrosive materials. For use to at least 13.6 Kg internal pressure. An ultimate vacuum of 10-7 can be realized with the standard three seals. Smooth acting valve permits fine adjustment of the opening. Front ring seal makes a positive closure against a precision-formed heavy glass seal. NOT bakeable. O-Rings are NOT replaceable. Reference marks on the stopcock body aid in repetitive setting. Replacement plugs with O-Rings sold separately.