Learn More
Ace Glass, Inc. 0-5mm vacuum stopcock w/UHDPE handle PTFE plug
SDP
Supplier:  Ace Glass, Inc. 8192207
Encompass_Preferred
0-5mm threaded vacuum stopcock with PTFE plug with UHDPE handle. Needle valve permits smooth valve adjustment down to 0.1 cc/min flow rate. Double PTFE ring seals prevent exposure of FETFE backup O-ring to corrosive gases and liquids.
Catalog No. 50-976-155