Learn More
Ace Glass, Inc. 0-5mm vacuum stopcock easy action
SDP
Catalog No. 50976154
Encompass_Preferred
0-5mm threaded vacuum stopcock with easy action PTFE plug with hooded, bakeable handle. Stopcock threads into a heavy wall glass barrel, permitting smooth adjustment. Double PTFE ring seals prevent exposure of FETFE backup O-ring to corrosive gases and liquids. Hooded handle can be removed from plug, making plug bakeable.
Catalog No. 50-976-154 Supplier Ace Glass, Inc. Supplier No. 819204
0-5mm threaded vacuum stopcock with easy action PTFE plug with hooded, bakeable handle. Stopcock threads into a heavy wall glass barrel, permitting smooth adjustment. Double PTFE ring seals prevent exposure of FETFE backup O-ring to corrosive gases and liquids. Hooded handle can be removed from plug, making plug bakeable.