Learn More
GFS Chemicals Inc CUPRIETHYLENEDIAMINE SOLUTION, 0.5M | 1L | GFS CHEMICALS, INC. Item #: 32112 | Bulk #: 6661
SDP
Catalog No. NC9928794
Encompass_Preferred
CUPRIETHYLENEDIAMINE SOLUTION, 0.5M | Chemical Formula : Cu(C!2H!8N!2)!2(OH)!2 | CAS # : 14552-35-3 | Specifications : Concentration (M) 0.49 - 0.51 M, Ratio En to Cu = 1.96 - 2.04
Catalog No. NC9928794 Supplier GFS Chemicals Inc Supplier No. 32112
Add to Cart
Add to Cart
CUPRIETHYLENEDIAMINE SOLUTION, 0.5M | Chemical Formula : Cu(C!2H!8N!2)!2(OH)!2 | CAS # : 14552-35-3 | Specifications : Concentration (M) 0.49 - 0.51 M, Ratio En to Cu = 1.96 - 2.04