Learn More
Boston Bioproducts Inc Saponin Cell Permeating Solution (0.5% w/v in PBS) - 4x100ml
Catalog No. 5000330337
Encompass_Preferred
Saponin Cell Permeating Solution (0 5% w/v in PBS) - 4x100ml (For Research Use and Further Manufacturing Only)
Catalog No. 50-003-30337 Supplier Boston Bioproducts Inc Supplier No. CLA1204X100ML
Saponin Cell Permeating Solution (0 5% w/v in PBS) - 4x100ml (For Research Use and Further Manufacturing Only)