Learn More
Laxco 0.5X Camera Tube Adjustable C-mount Interface for Z200 Series Stereo Microscope
GREENER_CHOICESDP
Catalog No. 12563728 Shop All Laxco Inc Products
Change view
Click to view available options
Type:
Camera Adapter
Catalog No. Type
12-563-728 Camera Adapter
1 options
Catalog No. 12-563-728 Supplier Laxco Supplier No. LMSPCTV0.5X
Add to Cart
Add to Cart
Enables users to attach any LAXCO SeBaCam camera using this accesory. Gives users to upgrade to documentation ability on their LAXCO stereo microscpe.
  • This product is made of premium material, durable in use and long life.
  • C‐mount adapter lens, mainly used to connect CCD camera or digital eyepiece to the microscope.
  • 0.5X magnification, which makes it easier to observe small objects more clearly and better observation effect.
  • Well‐made, easy to install and use.
  • Microscope accessories works best with LAXCO Z200 stereo microscope series
Specifications
Type Camera Adapter
Sterility Sterile