Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Rad Laboratories 0.6 μm Gold Microcarriers, 500 Pack, 0.25 g, Compatible with PDS-1000/He and Helios gene gun systems, for biolistic transformation

Catalog No. NC1775193
Encompass

The Bio-Rad Laboratories Inc. 0.6 μm Gold Microcarriers are expertly designed for effective gene delivery in biolistic transformation systems. With a diameter of 0.6 μm, these microcarriers are ideal for use in instruments like the PDS-1000/He and Helios gene gun systems. The gold material ensures high density and chemical stability, making it a reliable option for delivering DNA or other biomolecules into cells. Each pack includes 0.25 g of microcarriers, enabling consistent experimental outcomes essential for scientific research that demands accurate gene transfer.

  • Material: Gold
  • Diameter: 0.6 μm
  • Quantity: 0.25 g
  • Compatibility: PDS-1000/He gene gun system
  • Compatibility: Helios gene gun system
  • Application: Biolistic transformation

Catalog No. NC1775193 Supplier Bio Rad Laboratories Supplier No. 1652262
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Bio-Rad Laboratories Inc. 0.6 μm Gold Microcarriers are expertly designed for effective gene delivery in biolistic transformation systems. With a diameter of 0.6 μm, these microcarriers are ideal for use in instruments like the PDS-1000/He and Helios gene gun systems. The gold material ensures high density and chemical stability, making it a reliable option for delivering DNA or other biomolecules into cells. Each pack includes 0.25 g of microcarriers, enabling consistent experimental outcomes essential for scientific research that demands accurate gene transfer.

  • Material: Gold
  • Diameter: 0.6 μm
  • Quantity: 0.25 g
  • Compatibility: PDS-1000/He gene gun system
  • Compatibility: Helios gene gun system
  • Application: Biolistic transformation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.