Learn More
Ace Glass, Inc. Gauge, pressure, 0-60psi, stainless steel internal materials, 1/8in male NPT bottom fitting, 1.5in diameter
SDP
Catalog No. 50983494
Encompass
0-60psi pressure gauge with stainless steel internal materials. Features 1/8 male NPT bottom fitting. Gauges diameter is 1.5 inches.
Catalog No. 50-983-494 Supplier Ace Glass, Inc. Supplier No. 1338544
0-60psi pressure gauge with stainless steel internal materials. Features 1/8 male NPT bottom fitting. Gauges diameter is 1.5 inches.