Learn More
Andwin Scientific 0.65 TUBES PK/1000
SDP
Catalog No. NC9260727
Encompass_Preferred
The GeneMate 0.65 mL Graduated Microcentrifuge Tubes are ideal for boiling procedures. This product is autoclavable and can operate within the temperature range of -90C to 120C. It has a positive flat cap seal design that will not open snap seal during boiling. The cap also makes it easier to repeatedly open and close the tubes. The tubes have frosted writing surfaces on the cap and side. The graduations are at every 100 L. This is sold in a pack of 1000.
Catalog No. NC9260727 Supplier Andwin Scientific Supplier No. C32591
Add to Cart
Add to Cart
The GeneMate 0.65 mL Graduated Microcentrifuge Tubes are ideal for boiling procedures. This product is autoclavable and can operate within the temperature range of -90C to 120C. It has a positive flat cap seal design that will not open snap seal during boiling. The cap also makes it easier to repeatedly open and close the tubes. The tubes have frosted writing surfaces on the cap and side. The graduations are at every 100 L. This is sold in a pack of 1000.