Learn More
Diagenode Inc 0.65 ml BioruptorPico Microtubes 500 tubes.
Catalog No. NC1062171
Encompass
The 0.65 ml Bioruptor Microtubes for DNA and chromatin shearing strongly improve DNA and chromatin sonication efficiency. These tubes have been thoroughly validated for use on the Bioruptor Pico (B0106000) or Bioruptor NGS (UCD-600) and are strongly recommended for DNA and chromatin shearing applications using these instruments.Recommended sample volume: 100 l.
Catalog No. NC1062171 Supplier Diagenode Inc Supplier No. C30010011
The 0.65 ml Bioruptor Microtubes for DNA and chromatin shearing strongly improve DNA and chromatin sonication efficiency. These tubes have been thoroughly validated for use on the Bioruptor Pico (B0106000) or Bioruptor NGS (UCD-600) and are strongly recommended for DNA and chromatin shearing applications using these instruments.Recommended sample volume: 100 l.