Learn More
ACCU-SCOPE™ 0.65x C-Mount Adapter
SDP
Catalog No. 01189299 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Type:
Camera Adapter
Catalog No. Type
01-189-299 Camera Adapter
1 options
Catalog No. 01-189-299 Supplier ACCU-SCOPE™ Supplier No. 00201065
Add to Cart
Add to Cart
Microscope camera 0.65x c-mount adapter for 0.65 in. chip sensors with adjustable focus. Adapter can be focused to match the focus of the digital image to the image seen through the eyepieces. Ideal for use with 0.5 in. diagonal camera sensors or smaller.
  • 0.65x c-mount adapter to attach camera to EXC-400, EXC-350, 3001-LED or 3001 microscopes.
  • Adapter can be focused to match the focus of the digital image to the image seen through the eyepieces.
  • Ideal for use with 0.65 in. diagonal camera sensors or smaller.
Specifications
Type Camera Adapter
Autoclavable Not Autoclavable
Sterility Non-sterile