Learn More
Chemglass Life Sciences 0.6mL Micro Centrifuge Tube, Clear, Conical, Lock Cap, Sterile (50/Bag, 750/Pk)
SDP
Catalog No. 501897710
Encompass_Preferred
• Sterile • USP VI grade polypropylene material, free from heavy metal • RCF: 30,000 x g • Endotoxin level: <0.1EU • Clear graduations and frosted marking area • Special Safe-lock cap design provides secure lid sealing • Easy opening and closing • Temperature range: -80℃ to +121℃ • DNase/RNase-freeTubes are autoclavable at 121°C (250°F), under 15psi for 10–15 minutes with drying. Remove from autoclave immediately. Excessive exposure to autoclaving heat may cause warping or decreased structural integrity.
Catalog No. 50-189-7710 Supplier Chemglass Life Sciences Supplier No. CGN430006N
• Sterile • USP VI grade polypropylene material, free from heavy metal • RCF: 30,000 x g • Endotoxin level: <0.1EU • Clear graduations and frosted marking area • Special Safe-lock cap design provides secure lid sealing • Easy opening and closing • Temperature range: -80℃ to +121℃ • DNase/RNase-freeTubes are autoclavable at 121°C (250°F), under 15psi for 10–15 minutes with drying. Remove from autoclave immediately. Excessive exposure to autoclaving heat may cause warping or decreased structural integrity.