Learn More
BioSpec Products 0.7 mm Zirconia Beads 1lb bottle
SDP
Catalog No. NC0418963
Encompass_Preferred
Zirconia beads have a density of 5.5g/cc (Twice as dense as glass beads - good for tough tissue).  Listed bead sizes are median (average) values. Because beads are sorted by sieves, bead size varies between +/-10% of a listed value. 1 lb bottle
Catalog No. NC0418963 Supplier BioSpec Products Supplier No. 11079107ZX
Zirconia beads have a density of 5.5g/cc (Twice as dense as glass beads - good for tough tissue).  Listed bead sizes are median (average) values. Because beads are sorted by sieves, bead size varies between +/-10% of a listed value. 1 lb bottle