Learn More
USA Scientific Inc 0.75 mL Screw Cap Tubes, v-bot
Catalog No. 502745022
Encompass
0.75mL 2D coded Screw Cap tubes V-bottom in Micronic 96-2 Rack with low cover (Barcoded A1-H1 side), 10 racks
Catalog No. 50-274-5022 Supplier USA Scientific Inc Supplier No. 17752519
0.75mL 2D coded Screw Cap tubes V-bottom in Micronic 96-2 Rack with low cover (Barcoded A1-H1 side), 10 racks