Learn More
Alkali Scientific 0.75mL 11mm PP Snap Top Micro Vial Transparent Conical Bottom. 12x32mm, 100/Pk
SDP
Catalog No. 5000663051
Encompass_Preferred
The Alkali Scientific™ 0.75 mL PP Snap Top Micro Vial is designed for secure storage and precise handling of small volume samples in laboratory and analytical workflows. Made from durable transparent polypropylene (PP), this vial offers excellent chemical resistance and full visibility of contents. The conical bottom design ensures maximum sample recovery and minimizes residual volume, while the snap top closure provides a reliable seal without the need for additional caps. Measuring 12 × 32 mm, it is ideal for autosamplers, chromatography, and routine laboratory use. Supplied in packs of 100, these micro vials deliver consistent performance for high throughput applications. Key Features 0.75 mL capacity for precise small volume sample handling Made from durable transparent polypropylene (PP) with excellent chemical resistance Conical bottom design maximizes sample recovery and minimizes residual volume Snap top closure provides a secure seal without separate c
Catalog No. 50-006-63051 Supplier Alkali Scientific Supplier No. MVP07.5
The Alkali Scientific™ 0.75 mL PP Snap Top Micro Vial is designed for secure storage and precise handling of small volume samples in laboratory and analytical workflows. Made from durable transparent polypropylene (PP), this vial offers excellent chemical resistance and full visibility of contents. The conical bottom design ensures maximum sample recovery and minimizes residual volume, while the snap top closure provides a reliable seal without the need for additional caps. Measuring 12 × 32 mm, it is ideal for autosamplers, chromatography, and routine laboratory use. Supplied in packs of 100, these micro vials deliver consistent performance for high throughput applications. Key Features 0.75 mL capacity for precise small volume sample handling Made from durable transparent polypropylene (PP) with excellent chemical resistance Conical bottom design maximizes sample recovery and minimizes residual volume Snap top closure provides a secure seal without separate c