Learn More
JDM LAB SOLUTIONS LLC 0.75ml external thread 2D Tube
Supplier:  JDM LAB SOLUTIONS LLC 2010801
Encompass
0 75ml 2D barcoded external thread storage tube SBS rack format precapped with screw cap sterile
Catalog No. 50-000-14264