Learn More
JDM LAB SOLUTIONS LLC 0.75ml external thread 2D Tube
Supplier:  JDM LAB SOLUTIONS LLC 201086
Encompass
0 75ml 2D barcoded external thread storage tube no cap Amber AlteCleanMinimum order quantity 15 cases
Catalog No. 50-000-14299