Learn More
JDM LAB SOLUTIONS LLC 0.75ml external thread 2D Tube
Supplier:  JDM LAB SOLUTIONS LLC 201909
Encompass
0 75ml 2D barcoded external thread storage tube with 1D linear barcode human readable tube ID SBS rack format pre-capped with screw cap AlteClean
Catalog No. 50-000-14275