Learn More
JDM LAB SOLUTIONS LLC 0.75ml external thread 2D Tube
Supplier:  JDM LAB SOLUTIONS LLC 2010971
Encompass
0 75ml 2D barcoded external thread storage tube SBS rack format precapped with GREY screw cap sterile
Catalog No. 50-000-14235