Learn More
JDM LAB SOLUTIONS LLC 0.75ml Internal thread 2D Tube
Supplier:  JDM LAB SOLUTIONS LLC 201018
Encompass
0 75ml 2D barcoded internal thread storage tube SBS rack format no cap AlteClean
Catalog No. 50-000-14311