Learn More
JDM LAB SOLUTIONS LLC 0.75ml Internal thread 2D Tube
Supplier:  JDM LAB SOLUTIONS LLC 2010261
Encompass
0 75ml 2D barcoded internal thread storage tube SBS rack format precapped with BLUE screw cap sterile
Catalog No. 50-000-14265