Learn More
JDM LAB SOLUTIONS LLC 0.75ml no thread 2D Tube
Supplier:  JDM LAB SOLUTIONS LLC 2010291
Encompass
0 75ml 2D barcoded no thread storage tube SBS rack format pre-capped with natural TPE septum cap sterile
Catalog No. 50-000-14206