Learn More
Analytical Sales & Services 0.7mL Tapered/Flat-Bottom Clear Glass Inserts in 96-Well Plate with Pierceable "X" Cap Mat
Catalog No. 5000804411
Encompass
0.7mL Volume Flat Bottom Tapered Glass Inserts in 96-Well Sample Prep Plate\n\n• Comes with 2mL polypropylene pierceable Cap Mat with "X" on surface of each well position\n\n• Pre-assembled\n\nQty/Pkg: EACH
Catalog No. 50-008-04411 Supplier Analytical Sales & Services Supplier No. 96326
0.7mL Volume Flat Bottom Tapered Glass Inserts in 96-Well Sample Prep Plate\n\n• Comes with 2mL polypropylene pierceable Cap Mat with "X" on surface of each well position\n\n• Pre-assembled\n\nQty/Pkg: EACH