Learn More
Analytical Sales & Services 0.7mL Tapered/Flat-Bottom Clear Glass Inserts in 96-Well Plate with Pre-Slit Silicone Clear Silicone Ultra Thin Cap Mat
Catalog No. 5000804336
Encompass
0.7mL Volume Flat Bottom Tapered Glass Inserts in 96-Well Sample Prep Plate\n\n• Comes with Pre-Slit Silicone Cap Mat\n\n• Pre-Assembled\n\nQty/Pkg: EACH
Catalog No. 50-008-04336 Supplier Analytical Sales & Services Supplier No. 96380
0.7mL Volume Flat Bottom Tapered Glass Inserts in 96-Well Sample Prep Plate\n\n• Comes with Pre-Slit Silicone Cap Mat\n\n• Pre-Assembled\n\nQty/Pkg: EACH