Learn More
Analytical Sales & Services 0.7mL Tapered/Flat-Bottom Clear Glass Inserts in Vial Loader
Supplier:  Analytical Sales & Services 96305VL
Encompass
0.7mL Clear Glass Tapered, Flat-Bottom Inserts in Vial Loader\n\n• Vial loader contains 96 inserts\n\nQty/Pkg: Each
Catalog No. 50-008-06077