Learn More
Analytical Sales & Services 0.7mL Tapered/Round-Bottom Clear Glass Inserts in 96-Well Plate with Pierceable Cap Mat - SILANIZED
Catalog No. 5000806079
Encompass
0.7mL Tapered Glass Inserts in 96-Well Sample Prep Plate\n\n• Silanized Glass Vials\n• Comes with Pierceable Polypropylene Cap Mat\n• Pre-assembled\n\nQty/Pkg: EACH
Catalog No. 50-008-06079 Supplier Analytical Sales & Services Supplier No. 96627S
0.7mL Tapered Glass Inserts in 96-Well Sample Prep Plate\n\n• Silanized Glass Vials\n• Comes with Pierceable Polypropylene Cap Mat\n• Pre-assembled\n\nQty/Pkg: EACH