Learn More
Analytical Sales & Services 0.7mL Tapered/Round-Bottom Clear Glass Inserts in Vial Loader
Catalog No. 5000806450
Encompass
0.7mL Clear Glass Tapered, Round-Bottom Inserts in Vial Loader\n\n• Vial loader contains 96 inserts\n\nQty/Pkg: Each\n\n 
Catalog No. 50-008-06450 Supplier Analytical Sales & Services Supplier No. 96837VL
Add to Cart
Add to Cart
0.7mL Clear Glass Tapered, Round-Bottom Inserts in Vial Loader\n\n• Vial loader contains 96 inserts\n\nQty/Pkg: Each\n\n 