Learn More
Chemglass Life Sciences 0-8mm Chem-Vac, Chem-Cap Valve Replacement Plug and Control Knob, High Vacuum
SDP
Catalog No. CG96102
Encompass_Preferred
0-8mm Chem-Vac, Chem-Cap Valve Replacement Plug and Control Knob, High Vacuum. Replacement CHEM-CAP plug and control knob for CHEM-VAC high vacuum valves CG-951 through CG-960. Plug is sold complete with control knob and CHEM-CAP stem.
Catalog No. CG96102 Supplier Chemglass Life Sciences Supplier No. CG96102
0-8mm Chem-Vac, Chem-Cap Valve Replacement Plug and Control Knob, High Vacuum. Replacement CHEM-CAP plug and control knob for CHEM-VAC high vacuum valves CG-951 through CG-960. Plug is sold complete with control knob and CHEM-CAP stem.