Learn More
Chemglass Life Sciences 0-8mm Chem-Vac Valve Replacement Plug and Control Knob, TFE, Viton O-Ring, High Vacuum
SDP
Catalog No. CG93502
Encompass_Preferred
0-8mm Chem-Vac Valve Replacement Plug and Control Knob, TFE, Viton O-Ring, High Vacuum. Replacement TFE plug and control knob for CHEM-VAC high vacuum valves CG-925 through CG-934. Plug is sold complete with control knob and TFE stem having three o-rings.PLEASE NOTE: CONTROL KNOB IS NOT SOLD SEPARATELY ON THE CG-900 SERIES VALVES. THE 0-20mm BORE SIZE VALVE IS OUR PREVIOUS STYLE CG-500 SERIES VALVE WITH COARSE THREAD.
Catalog No. CG93502 Supplier Chemglass Life Sciences Supplier No. CG93502
0-8mm Chem-Vac Valve Replacement Plug and Control Knob, TFE, Viton O-Ring, High Vacuum. Replacement TFE plug and control knob for CHEM-VAC high vacuum valves CG-925 through CG-934. Plug is sold complete with control knob and TFE stem having three o-rings.PLEASE NOTE: CONTROL KNOB IS NOT SOLD SEPARATELY ON THE CG-900 SERIES VALVES. THE 0-20mm BORE SIZE VALVE IS OUR PREVIOUS STYLE CG-500 SERIES VALVE WITH COARSE THREAD.