Learn More
Hach Company 0.8 M Sodium Hydroxide Solution, 1 L (Lachat reagent for FIA)
Catalog No. NC3824195 Shop All Hach Company Products
Encompass
Ready-to-use 0.8 M sodium hydroxide reagent supplied in a 1 L bottle for use with flow injection analysis (FIA) ammonia determinations. The solution is prepared for laboratory analytical use and is shipped under hazardous materials regulations; consult the Safety Data Sheet (SDS) before handling or shipping.
  • Ready-to-use 0.8 M sodium hydroxide solution.
  • Supplied in a 1 L bottle.
  • Intended for ammonia analysis by flow injection analysis (FIA).
  • Pre-measured and quality-controlled for analytical consistency.
  • Hazardous material; consult SDS and follow appropriate handling and shipping procedures.
Catalog No. NC3824195 Supplier Hach Company Supplier No. 52008
Ready-to-use 0.8 M sodium hydroxide reagent supplied in a 1 L bottle for use with flow injection analysis (FIA) ammonia determinations. The solution is prepared for laboratory analytical use and is shipped under hazardous materials regulations; consult the Safety Data Sheet (SDS) before handling or shipping.
  • Ready-to-use 0.8 M sodium hydroxide solution.
  • Supplied in a 1 L bottle.
  • Intended for ammonia analysis by flow injection analysis (FIA).
  • Pre-measured and quality-controlled for analytical consistency.
  • Hazardous material; consult SDS and follow appropriate handling and shipping procedures.