Learn More
Embi Tec 0.8% TAE GEL NO EB 6 WELL
SDP
Catalog No. NC0106640
Encompass
0.8% Agarose TAE without EtBr, Medium Prep Gel, 6 well, 2.4mm thick comb, 5/bx
Catalog No. NC0106640 Supplier Embi Tec Supplier No. GE5181
0.8% Agarose TAE without EtBr, Medium Prep Gel, 6 well, 2.4mm thick comb, 5/bx