Learn More
MSC Dwyer MARK II WSI Wind Indicator, 0-80 mph, 1 ea
Catalog No. 5000504015
Encompass
The Series Mark II Wind Speed Indicator is a durable, molded white plastic instrument that indicates wind speed directly on a liquid-filled scale. It is suitable for various applications, including weather monitoring stations and general use in homes or offices.
  • Durable molded white instrument with gold scale
  • English and metric units available
  • Accurate, low-cost, and practical
  • Includes ABS plastic vane, 50 ft tubing, mounting hardware, and gage fluid
  • Measures wind speed from 0-80 mph or 0-130 km/h
Catalog No. 50-005-04015 Supplier MSC Supplier No. 04293270
The Series Mark II Wind Speed Indicator is a durable, molded white plastic instrument that indicates wind speed directly on a liquid-filled scale. It is suitable for various applications, including weather monitoring stations and general use in homes or offices.
  • Durable molded white instrument with gold scale
  • English and metric units available
  • Accurate, low-cost, and practical
  • Includes ABS plastic vane, 50 ft tubing, mounting hardware, and gage fluid
  • Measures wind speed from 0-80 mph or 0-130 km/h