Learn More
Chemglass Life Sciences 0-8mm Chem-Vac Valve, Three Way, Glass Portion Only, 19mm OD Barrel, 12.7mm OD x 1.6 W Sidearms
SDP
Catalog No. 501531869
Encompass_Preferred
0-8mm Chem-Vac Valve, Three Way, Glass Portion Only, 19mm OD Barrel, 12.7mm OD x 1.6 W Sidearms. CHEM-VAC valve glass portion only. Three-way configuration for use in construction of manifolds, adapters, etc. Valve has two barrels sealed together 180 from one another, each having a sidearm of the listed size and a common arm sealed between the barrels. Heavy duty rounded coarse thread and precision formed tapered seat.
Catalog No. 50-153-1869 Supplier Chemglass Life Sciences Supplier No. CG90602
0-8mm Chem-Vac Valve, Three Way, Glass Portion Only, 19mm OD Barrel, 12.7mm OD x 1.6 W Sidearms. CHEM-VAC valve glass portion only. Three-way configuration for use in construction of manifolds, adapters, etc. Valve has two barrels sealed together 180 from one another, each having a sidearm of the listed size and a common arm sealed between the barrels. Heavy duty rounded coarse thread and precision formed tapered seat.