Learn More
Chemglass Life Sciences Adapter, Connecting, Inlet, Airfree, Schlenk, 0-8mm Valve
SDP
Catalog No. AF007402
Encompass_Preferred
Adapter, Connecting, Inlet, Airfree, Schlenk, 0-8mm Valve. Similar to the AF-0072 adapter, but with two #15 o-ring joints. Available in 0-4mm, 0-8mm or 0-12mm valve sizes. Each adapter is supplied complete with (2) #116 Viton o-rings.
Catalog No. AF007402 Supplier Chemglass Life Sciences Supplier No. AF007402
Adapter, Connecting, Inlet, Airfree, Schlenk, 0-8mm Valve. Similar to the AF-0072 adapter, but with two #15 o-ring joints. Available in 0-4mm, 0-8mm or 0-12mm valve sizes. Each adapter is supplied complete with (2) #116 Viton o-rings.