Learn More
Next Advance Inc 0.9-2MM BLND RNASE SS BEAD 4ML
SDP
Catalog No. NC0586331
Encompass
0.9 - 2.0 mm stainless steel bead blend. Rnase-free, density is 7.9 g / mL, 4mL package. For the homogenization of tough samples.
Catalog No. NC0586331 Supplier Next Advance Inc Supplier No. SSB14BRNA
0.9 - 2.0 mm stainless steel bead blend. Rnase-free, density is 7.9 g / mL, 4mL package. For the homogenization of tough samples.