Learn More
General Laboratory Products Saline (0.9%) - 100ml, 12 vials per case
Catalog No. NC0720237
Encompass_Preferred
Saline (0.9%) - 100ml. Saline is routinely used a diluent. This is an industrial use product, not for humans and not to be used as an injectable. 12 Vials per case. Used as a diluent.
Catalog No. NC0720237 Supplier General Laboratory Products Supplier No. 125EZ7002100SEP
Saline (0.9%) - 100ml. Saline is routinely used a diluent. This is an industrial use product, not for humans and not to be used as an injectable. 12 Vials per case. Used as a diluent.