Learn More
Thomas Scientific Cryo Laser Labels, 0.94" x 0.5", Removable, White
SDP
Catalog No. NC9436812
Encompass
Cryogenic plastic labels for long-term storage in liquid nitrogen and deep-freezing harsh environments for biomedical research and clinical laboratories. Cryo labels accept desktop laser printing and permanent marker(*). Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and Gamma irradiation. (*) RCL-12 labels are not recommended for using on tight circumference vials. Recommended for flat surfaces only.
Catalog No. NC9436812 Supplier Thomas Scientific Supplier No. 1189D61
Cryogenic plastic labels for long-term storage in liquid nitrogen and deep-freezing harsh environments for biomedical research and clinical laboratories. Cryo labels accept desktop laser printing and permanent marker(*). Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and Gamma irradiation. (*) RCL-12 labels are not recommended for using on tight circumference vials. Recommended for flat surfaces only.