Learn More
Ace Glass, Inc. Heating tape, 1in, 3ft length, 150w at 115v max, Glas-Col; 103A SET 13
SDP
Catalog No. 50977379
Encompass
1 Glas-Col heating tape allows the use of 115v without the need of a reducing voltage transformer. 3 foot length. Can be operated directly through a temperature control at normal outlet voltage. 150w at 115v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.
Catalog No. 50-977-379 Supplier Ace Glass, Inc. Supplier No. 1206342
1 Glas-Col heating tape allows the use of 115v without the need of a reducing voltage transformer. 3 foot length. Can be operated directly through a temperature control at normal outlet voltage. 150w at 115v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.