Learn More
MSC Mitutoyo Electronic Outside Micrometer, 1.2 in, Solid Carbide, 1 ea
Catalog No. NC1963413
Encompass
This electronic outside micrometer is designed for precise external measurements, offering a range from 0.00 mm to 1.2 in. It features a solid carbide measuring face and an IP65 rating for protection against dust and water.
  • Resolution of 0.000050 in
  • Accuracy of ±0.000100 in
  • Friction ratchet thimble for consistent measuring force
  • IP65 rated for harsh environments
  • Includes a plastic case for protection
  • TUV certified for quality standards
  • SR44 battery included
Catalog No. NC1963413 Supplier MSC Supplier No. 06385421
This electronic outside micrometer is designed for precise external measurements, offering a range from 0.00 mm to 1.2 in. It features a solid carbide measuring face and an IP65 rating for protection against dust and water.
  • Resolution of 0.000050 in
  • Accuracy of ±0.000100 in
  • Friction ratchet thimble for consistent measuring force
  • IP65 rated for harsh environments
  • Includes a plastic case for protection
  • TUV certified for quality standards
  • SR44 battery included