Learn More
Polysciences, Inc. JB-4 Embedding Kit 1 kit Contents of kit: 800ml solution A; 30ml solution B; 12g catalyst
SDP
Catalog No. NC9369351
Encompass_Preferred
JB-4 Embedding Kit 1 kit Contents of kit: 800ml solution A; 30ml solution B; 12g catalyst Use in the preparation of embedded samples for high resolution light microscopy Handling: Glove chemical goggles and chemical mask or hood
Catalog No. NC9369351 Supplier Polysciences, Inc. Supplier No. 002261
JB-4 Embedding Kit 1 kit Contents of kit: 800ml solution A; 30ml solution B; 12g catalyst Use in the preparation of embedded samples for high resolution light microscopy Handling: Glove chemical goggles and chemical mask or hood